Drukuj
Systemy filtracji próżniowej na górze butelki, membrana nylonowa, Rapid-Flow™ Nalgene®
Dostawca: Thermo Fisher Scientific
150-0020 150-0045 151-4020 151-4045 153-0020 153-0045 154-0020 154-0045 163-0020 164-0020
734-5012EA 1180 PLN
734-5012 734-5013 734-5014 734-5015 734-5016 734-5017 734-5018 734-5019 513-1631 513-1641
Systemy filtracji próżniowej na górze butelki, membrana nylonowa, Rapid-Flow™ Nalgene®
Filtry Zestawy do filtracji, plastikowe
Rapid-Flow™ disposable filter units feature the exclusive rapid-flow support column design, providing fast flow rates and high throughput.
  • Color coded according to membrane
  • Fastest flow rate and lowest clogging of comparable alternatives
  • 0.1 µm PES available for mycoplasma protection
  • 0.2 µm PES devices are stem cell and mouse embryonic assay (MEA) tested
Ideal for filtration of cell culture media, serum, additives and buffers.
Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum
Więcej informacji o produkcie