Drukuj
Węże do dializy, zestawy próbne
Dostawca: Spectrum Laboratories
131486T 131414T 131054T 131090T 131198T 131270T 131342T 131378T 131450T 132720T 132655T 132678T 132665T 132680T 132725T 132676T 132754T 132724T 132709T 132703T 132682T 132675T 132670T 132660T 132650T 132645T 132684T
734-2453EA 1310 PLN
734-2453 734-2451 734-2445 734-2446 734-2447 734-2448 734-2449 734-2450 734-2452 734-2443 734-2439 734-2441 734-2440 734-2442 734-2444 734-2640 734-2646 734-2643 734-2645 734-2644 734-2641 734-2639 734-2638 734-2637 734-2636 734-2635 734-2642
Węże do dializy, zestawy próbne
Przewody, kapilary i rury Rurki do dializy
Spectra/Por® tubing trial kits are available in 1 and 5 m lengths. They are an excellent choice for applications that require only a small amount of dialysis tubing.
  • Hydrophilic, symmetric porosity
Spectra/Por® 1, 2, 3, 4 and 5 come in two different tubing diameters and 1 or 5 m lengths for trial kits with a Biotech regenerated cellulose (RC) membrane. This covers all cut offs in the Spectra/Por® RC dialysis tubing range: 3,5 kDa, 6 - 8 kDa and 12 - 14 kDa. They contain one standard (for buoyance) and one weighted closure (for orientation).