Drukuj
Płytki do hodowli komórkowych, Falcon®
Dostawca: Corning
353502 353503 353504 355467
734-0065EA 1040 PLN
734-0065 734-0066 734-0067 734-1095
Płytki do hodowli komórkowych, Falcon®
Mikropłytki Płytki z insertami i inserty
PS, tissue culture-treated, sterile, non-pyrogenic, with lid. Specifically designed for use with Falcon® or Corning® BioCoat™ cell culture inserts, so that evaporation and contamination due to improper lid fit is eliminated.
  • Reagents can be added quickly and consistently for timed experiments
  • Aspiration from the well is easier, reducing the risk of contamination
  • Media cannot wick up between the insert and the well wall
  • Low-evaporation lid reduces evaporation and contamination
In the 'Feeding Position' pipette access is improved for fluid handling onWięcej informacji o produkcie