Drukuj
Płytki z poli-D-lizyną, Corning® BioCoat®
Dostawca: Corning
356509 354413 354414 354470 354509 356413 356414 356470
734-0298EA 3520 PLN
734-0298 734-0120 734-0121 734-0151 734-0174 734-0278 734-0279 734-0286
Płytki z poli-D-lizyną, Corning® BioCoat®
Mikropłytki Płytki do hodowli komórkowych
Poly-D-Lysine (PDL) and Poly-L-Lysine (PLL) are synthetic compounds that enhance cell adhesion and protein absorption by altering surface charges on the culture substrate. In addition to promoting cell adhesion, poly-lysine surface treatments support neurite outgrowth and improve the survival of many central nervous system (CNS) primary cells in culture. As PDL and PLL are synthetic molecules, they do not stimulate biological activity in the cells cultured on them, and they do not introduce impurities carried by natural polymers.
  • Attachment and spreading of a variety of cell types
  • Cell
Więcej informacji o produkcie