Drukuj
VWR®, Taq Plus 2X master mixes
Dostawca: VWR Chemicals
733-2599EA 3630 PLN
733-2599 733-2600 733-2597 733-2598
VWR®, Taq Plus 2X master mixes
Odczynniki do kwasów nukleinowych Zestawy do end-point PCR
VWR® Taq Plus is an optimised format of Taq DNA polymerase master mix and, therefore, is a good alternative to Taq DNA polymerase and Taq DNA polymerase master mix.
  • Ready to use 2X master mix for greater convenience
  • Significantly reduced set-up time
  • Increased specificity compared to Taq DNA polymerase master mix
  • Reduced risk of contamination
Ensures increased specificity and improved PCR performance, even on difficult templates. These reagents are suitable for standard PCR applications, screening and high throughput testing.
This PCR master mix is composed of Taq DNA polymerase and an optimised buffer system, including dNTPs and magnesium chloride (1,5 mM).
Zobacz ceny
SPECYFIKACJE
Opis Taq Plus 2x Master Mix 1.5mM MgCl2 2500 reactions Taq Plus 2x Master Mix 1.5mM MgCl2 5000 reactions Taq Plus 2x Master Mix 1.5mM MgCl2 100 reactions Taq Plus 2x Master Mix 1.5mM MgCl2 500 reactions
Environmentally Preferable
Format Mastermix incl. dNTPs Mastermix incl. dNTPs Mastermix incl. dNTPs Mastermix incl. dNTPs
Gorący start No No No No
Stężenie 2x 2x 2x 2x
Jednostki 2500 5000 100 500
Rozmiar 50 x 1.25 ml 25 x 5 ml 2 x 1.25 ml 10 x 1.25 ml
Wysoka wierność Nie Nie Nie Nie
Rozmiar amplikonu ≤ 4kb ≤ 4kb ≤ 4kb ≤ 4kb
Processivity Standard Standard Standard Standard
Aktywność egzonukleazy 5’→3’ 5’→3’ 5’→3’ 5’→3’
Product overhang 3'A 3'A 3'A 3'A
Warunki przechowywania – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months) – 20 °C (long term), + 4 °C (6 months)
Aplikacja Standard amplification Standard amplification Standard amplification Standard amplification