Drukuj
System dokumentacji żeli, Syngene G:BOX Chemi XRQ
Dostawca: Syngene (a division of the Synoptics group)
GBOX-CHEMI-XRQ
733-2350EA 89010 PLN
733-2350
System dokumentacji żeli, Syngene G:BOX Chemi XRQ
Systemy do fotodokumentacji
The Syngene G:BOX Chemi XRQ gel imaging system is designed for extended fluorescence applications and chemiluminescent Western blots. The G:BOX Chemi XRQ offers high quantum efficiency (73% at 425 nm) and 4 megapixel images with enhanced cooling for longer exposure times. Functional control of the G:BOX Chemi XRQ is handled by the GeneSys automatic control software, although full manual control is also possible.
  • Effective resolution up to 16 megapixels
  • Motor driven lens (with feedback) and seven position filter wheel (with UV filter to extend applications)
Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable
S×G×W 570×450×840 mm