Drukuj
Agaroza, SeaKem® Genetic Technology Grade™ (GTG)
Dostawca: LONZA
50074 50070 50071
733-1542EA 10280 PLN
733-1542 733-1540 733-1541
Agaroza, SeaKem® Genetic Technology Grade™ (GTG)
Elektroforezy, Żele Agaroza i akrylamid
SeaKem® is a high gel strength, low EEO, standard gelling temperature agarose perfect for electrophoresis and other molecular biology applications.
  • Quality tested to certify performance
  • High gel strength
  • Low EEO
  • Standard melting temperature
This Genetic Technology Grade™ (GTG) agarose is specifically designed for preparative DNA electrophoresis and is tested for the presence of proteases, ligases and nucleases. DNA recovered from SeaKem® GTG™ Agarose gels may be reliably digested and ligated. SeaKem® GTG™ Agarose is extensively performance tested toWięcej informacji o produkcie