Drukuj
Wieczka wypukłe w paskach
Dostawca: Thermo Fisher Scientific
AB-0386 AB-0602 AB-0852 AB-0853 AB-0265
732-4806EA 233 PLN
732-4806 732-4830 732-4898 732-4899 732-4801
Wieczka wypukłe w paskach
Probówki Probówki do PCR
Suitable for use with 96-well PCR plates as well as standard and low profile tube strips.
  • Made from Polypropylene (PP)
  • Large end tabs for strip removal
  • Certified free from DNase, RNase and human DNA