Drukuj
SE250 Mighty Small II Mini urządzenie do elektroforezy pionowej
Dostawca: HOEFER ELECTROPHORESIS
SE250-10A-.75
732-1752EA 4730 PLN
732-1752
SE250 Mighty Small II Mini urządzenie do elektroforezy pionowej
Systemy do elektroforezy Systemy do elektroforezy pionowej
The SE250 Mighty Small II mini vertical protein electrophoresis unit is designed for quick screening and rapid electrophoretic separation.
  • Maximum glass plate size 10×8 cm
  • Efficient active cooling ensures sharp bands
  • Quick and easy to assemble
  • Requires minimal buffer
Easily upgradable to the SE260 (with optional accessories) resulting in the additional capability of accommodating both 10×8 and 10×10,5 cm gels, enabling a wider variety of precast gel options.
Dodatkowe informacje: Complete system includes lower buffer chamber, upper buffer chamber/Więcej informacji o produkcie