Drukuj
Aparaty do elektroforezy poziomej, Edvotek M6 i M12
Dostawca: EDVOTEK
500 502 504
732-1417EA 1550 PLN
732-1417 732-1419 732-1420
Aparaty do elektroforezy poziomej, Edvotek M6 i M12
Systemy do elektroforezy Systemy do elektroforezy poziomej
Capable of running one or two groups of student samples at the same time and producing excellent results in 30 to 40 minutes.
  • Seamless, injection-moulded casing
  • Safety interlock cover
  • Corrosion-resistant platinum electrodes
  • Safety insulated electrical leads
  • Adjustable leveling feet