Drukuj
Lampy UV, PL Compact
Dostawca: UVP ULTRA VIOLET PRODUCTS
95-0021-10 95-0016-15 95-0018-03 95-0017-10 95-0019-03
732-0859EA 1850 PLN
732-0859 733-2366 730-1525 732-0858 730-1526
Lampy UV, PL Compact
Oświetlenie Lampy UV
Made of durable, lightweight plastic, these compact 4 W UV lamps can be used as a handheld instrument or hands-free in combination with an optional stand. Suitable for a wide range of applications, including fluorescence of stained gels and sterilisation. UV tubes and filters can be replaced simply and conveniently.
  • Lightweight, easy to handle lamp design
  • On/off button conveniently located on the back of the lamp
  • Models available include 254 nm shortwave, 365 nm longwave or a combination shortwave/longwave (365/254 nm)
  • Shortwave/longwave version (UVGL-15)
Więcej informacji o produkcie