Drukuj
Samoprzylepne foliowe uszczelnienie płytek i ultra czyste paski mocujące do qPCR
Dostawca: Thermo Fisher Scientific
AB-0866
732-0081EA 312 PLN
732-0081
Samoprzylepne foliowe uszczelnienie płytek i ultra czyste paski mocujące do qPCR
Probówki Probówki do PCR
PP flat 8-cap strips designed for qPCR and other fluorescence applications.
  • Compatible with all Thermo Scientific™ 96-well PCR plates, as well as standard and low profile tube strips
  • Specially formulated ultra clear polypropylene enhances signal detection by minimising autoflourescence and maximising transmission
  • Highly polished flat surface provides an optically clear window to deliver maximum and consistent signal transmission, critical for accurate qPCR results