Drukuj
Dilutor grawimetryczny, IUL Pinch Dilutor
Dostawca: IUL Instruments SA
7511
710-0862EA 8530 PLN
710-0862
Dilutor grawimetryczny, IUL Pinch Dilutor
Dilutory / urządzenia do rozcieńczania Dilutory grawimetryczne
Equipment specifically designed to facilitate the controlled addition of media into a homogeniser/lab blender bag using a load cell. The Pinch Dilutor constantly monitors the weight in the homogeniser bag and consequently distributes the precise quantity of diluent into the bag.
  • Ideal for dilution of solid and semi-solid samples
  • Simple to operate and control dilution with pinch valve
  • Smooth, silent and fast process
  • Contact parts and tubing can easily be dismantled and sterilised in an autoclave