Drukuj
Rybonukleaza A, bydlęca (z trzustki)
Dostawca: Merck Millipore (Novagen)
70856-3
70856-3EA 551 PLN
70856-3
Rybonukleaza A, bydlęca (z trzustki)
Enzymy
Convenient solution for selective degradation of RNA.
This is a convenient alternative to powdered RNase A. It is a highly purified preparation of bovine pancreatic ribonuclease A suitable for use in selective removal of RNA. It has been pretreated to remove DNase I and is suitable for RNA digestion in plasmid purification procedures.
Supplied at a concentration of 10 mg/ml in 10 mM Tris-HCl, 1 mM EDTA, 50% glycerol, pH 7,5.