Drukuj
Spektrofotometry UV/Vis, DR6000
Dostawca: Hach
LPV4419900011 LPV4419900002 LPV4419900001
705-0913EA 84780 PLN
705-0913 705-1161 705-1160
Spektrofotometry UV/Vis, DR6000
Spektrofotometry i fotometry Spektrofotometry UV/Vis
UV/Vis spectrophotometers with radio frequency (RFID) technology to ensure high speed wavelength scanning across the UV and visible spectrum.
  • Improved laboratory performance thanks to 240 available methods
  • Transmittance (%), absorbance and concentration, scanning
  • RFID technology allows sample traceability
  • Optimal data management and reliable results
  • Key methods are possible including COD, chlorine, nitrates and ammonia
  • High quality reagents formulated to ensure outstanding performance and results
  • Software Quality Assurance (SQA) for scheduling, documenting
Więcej informacji o produkcie