Drukuj
Spektrofotometr VIS, photoLab® 7100
Dostawca: WTW
250203
705-0921EA 28650 PLN
705-0921
Spektrofotometr VIS, photoLab® 7100
Spektrofotometry i fotometry Spektrofotometry UV/Vis
The pHotoLab® 7100 combines routine analysis with spectral analysis and pioneering procedure OptRF.
  • Brilliant colour screen operation guidance
  • Multi-step and multi-wavelength programming tools
  • photoLab® color + data management included
  • Routine analysis with barcode recognition and automatic measurement range adaption for all cuvette sizes
Certyfikaty: CIE-compliant and other colour measurement options with optional PC software package photoLab® color+Data spectral.
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable
Tryby pomiaru Absorbance, % transmission, concentration, spectrum, kinetics
Wyświetlacz Backlit 7” colour display for extensive graphical evaluation
Liczba metod Over 200 pre-programmed methods, 1000 user-defined methods, 20 profiles for kinetics and spectra, comprehensive programming options
System optyczny Monochromator with grating and step motor / reference beam
Wavelength accuracy ±0,5 nm
Wavelength range 320 - 1100 nm
Interfejs 2× USB, Ethernet
Źródło światła Tungsten halogen
Pamięć 5000 ; 500 spectra (300 − 900 nm); 400 kinetics with 150 values
Szybkość skanowania 700 − 2000 nm/minutes in 1 ; 2 ; 5 ; 10 nm steps of wavelength range
Szerokość pasma widmowego 4 nm
S×G×W 404×314×197 mm
Waga 4,5 kg