Drukuj
Zestawy komparatorów, wieloparametrowe do badania wody, Comparator 2000+
Dostawca: Lovibond
413550 413570 413580
705-0361EA 19370 PLN
705-0361 705-0787 705-0362
Zestawy komparatorów, wieloparametrowe do badania wody, Comparator 2000+
Kolorymetry Komparatory
Lovibond® Comparator system 2000+ is an extremely versatile, modular system for testing water. The battery operated portable lighting unit can be integrated to give standardised lighting conditions to enhance reproducibility.
  • Compact and portable
  • Simple to use yet uncompromising in terms of precision and reproducibility of results
Dodatkowe informacje: Comparator test kits are supplied as a complete system in a sturdy plastic case. Together with the Comparator 2000+ and test discs, each kit includes all the necessary cells, accessories and Lovibond®Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable