Drukuj

Anti-IgE Goat Polyclonal Antibody

Dostawca: Tonbo Biosciences
70-8024-M001
70-8024-M001EA 1300 PLN
70-8024-M001
Anti-IgE Goat Polyclonal Antibody
Przeciwciała
Goat anti-Human IgE (epsilon chain) - Affinity Pure, min x w/bovine, mouse or rabbit serum proteins

Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: ε chain
Host: Goat
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR