Drukuj
Refraktometr ręczny, OE 200
Dostawca: REICHERT
137528L0
635-2115EA 1640 PLN
635-2115
Refraktometr ręczny, OE 200
Refraktometry Refraktometry przenośne
This handheld refractometer for wine provides fast and accurate measurements of dissolved sugars when testing grapes, citrus fruits, fruit juices, soft drinks, melons and maple sap for the winemaking industry.