Drukuj
Automatyczny refraktometr cyfrowy, RX-7000?
Dostawca: Atago
3262
635-0148EA 41500 PLN
635-0148
Automatyczny refraktometr cyfrowy, RX-7000?
Refraktometry Refraktometry nastołowe
The RX-7000α is an automatic digital refractometer, which can set measurement temperature internally and measure the refractive index, °Brix or concentration of various liquids accurately and speedily.
The RX-7000α with the ability to measure high refractive index and at higher temperature is particularly suitable for oils and fats with high melting points.