Drukuj
Kolorymetry spektrofotometryczne, Lovibond® seria PFXi 880
Dostawca: Lovibond Tintometer
1378801 1378801H 1378802 1378802H 1378803 1378803H 1378804 1378804H 1378805 1378805H 1378806 1378806H
633-0222EA 63470 PLN
633-0222 633-0223 633-0224 633-0225 633-0226 633-0227 633-0228 633-0229 633-0230 633-0231 633-0232 633-0233
Kolorymetry spektrofotometryczne, Lovibond® seria PFXi 880
Kolorymetry Kolorymetry automatycznej transmitacji
A series of automatic spectrophotometric colorimeters for objective colour analysis of edible oils, chemicals, petrochemicals, fuels and other transparent products. The PFXi 880 instruments are rugged colorimeters with fabricated steel housings, which function equally well as QC instruments within the laboratory or on 24-hour operation in a production environment. The accuracy and repeatability of the colour data provided by these colorimeters ensures consistency in colour measurement, giving processors the confidence needed to make important decisions regarding raw materials andWięcej informacji o produkcie