Drukuj
System przechowywania prowadnic
Dostawca: KARTELL
945L1 946L1 947L1 948L1
631-1283EA 5950 PLN
631-1283 631-1284 631-1285 631-1286
System przechowywania prowadnic
Pudełka na szkiełka mikroskopowe
Made from high impact PMMA and steel, this slide storage system is designed for secure classification and storage of microscope slides.
  • Lightweight and functional
  • Durable steel frame for stability
  • Semi-transparent, ice coloured drawers provide an attractive finish
  • Transparent lids permit easy drawer viewing, while protecting slides from dust and foreign bodies
  • System holds up to 1300 slides per drawer, 5200 slides in total
The system is composed of 4 sliding drawers, a closing cover, and 4 fixed support feet. As an alternative, the system is alsoWięcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable