Drukuj
VWR®, kolorowe szkiełka adhezyjne
Dostawca: VWR Collection
631-1169EA 950 PLN
631-1169 631-1172 631-1173 631-1175 631-1176
VWR®, kolorowe szkiełka adhezyjne
Szkiełka mikroskopowe i nakrywkowe Szkiełka mikroskopowe adhezyjne
Microscope slides recommended for haematoxylin and eosin (H&E) stain and hybridisation on frozen sections in histopathology examination.
  • Smooth edges for operational safety
  • Sticky surface for stronger adhesion than with poly-l-lysine
  • Marking with conventional labelling system, pencils, marker pens and thermal transfer printers
Recommended for hematoxylin and eosin (H&E) stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
Sposób pakowania: 50 pieces per box. 50 boxes per case.