Drukuj
Pudełko na szkiełka mikroskopowe
Dostawca: GLASWARENFABRIK KARL HECHT
42703010 42702010 42701020 42701010
631-0353EA 99.9 PLN
631-0353 631-0352 HECH42701020 HECH42701010
Pudełko na szkiełka mikroskopowe
Pudełka na szkiełka
These slide boxes are designed for secure, dust-free storage of specimen slides with metal catch and hinges.
  • Numbered index in the lid
  • Cardboard with black wooden frame
  • Numbered plastic grooves: Anti-static and xylene-resistant
Suitable for the safe and protected storage of slides.