Drukuj
Kamery do dokumentacji mikroskopowej
Dostawca: LEICA MICROSYSTEMS
13613735 11547106 11547118 11547113
630-3195EA 0 PLN
630-3195 630-3198 630-3437 630-3458
Kamery do dokumentacji mikroskopowej
Kamery
Leica (brightfield) colour cameras provide outstanding colour fidelity due to state-of-the-art colour interpolation algorithms performed in the camera head. Leica fluorescence cameras provide high sensor sensitivity allowing short exposure times and, therefore, preventing photo bleaching, so actively protecting cells from any photo damage.
  • Brightfield documentation cameras
  • Even fine structural and colour details can be distinguished due to appropriate pixel sizes for every desired microscope magnification
  • High definition (HD) display directly on a monitor