Drukuj
Odwrócone mikroskopy rutynowe ze stałym stolikiem i opcjami pakietu Fluo, DM IL LED
Dostawca: LEICA MICROSYSTEMS
11521288 11506303 11521265_P1 11521265_P2 11521266_P1 11521281_P1
630-3111EA 33000 PLN
630-3111 630-3300 630-3462 630-3463 630-3464 630-3465
Odwrócone mikroskopy rutynowe ze stałym stolikiem i opcjami pakietu Fluo, DM IL LED
Mikroskopy
The Leica DM IL is a fluorescence-ready digital imaging solution designed for use in a variety of laboratory settings including cell culture, micromanipulation, immuno staining and routine live imaging.
  • Inverted laboratory microscope with LED illumination
  • High performance optics
  • Ergonomic design
  • 5 W white light LED illumination
  • 20 mm field of view
  • 4-fold revolving nosepiece with 25x075 objective thread
The DM IL LED inverted microscope lab package is a robust inverted microscope with LED illumination designed for years of use without the need toWięcej informacji o produkcie