The Leica DM IL is a fluorescence-ready digital imaging solution designed for use in a variety of laboratory settings including cell culture, micromanipulation, immuno staining and routine live imaging.
- Inverted laboratory microscope with LED illumination
- High performance optics
- Ergonomic design
- 5 W white light LED illumination
- 20 mm field of view
- 4-fold revolving nosepiece with 25x075 objective thread
The DM IL LED inverted microscope lab package is a robust inverted microscope with LED illumination designed for years of use without the need to replace bulbs.
The DM IL LED is fully configured for both brightfield and fluorescence imaging. It features an LED light source comprising of five solid state sources, operating simultaneously to produce up to 5 W of white light. The DM IL features long working objectives from 4× to 40×. Objectives 10×, 20× and 40× each have phase rings inscribed on them that are matched to the Phase-1, Phase-2 and Phase-3; light rings contained within a single slider. The DM IL is available either with a camera port or stand-alone. When properly configured, the DM IL can be combined with a selection of cameras from the Leica camera portfolio. Additional stage accessories are available to fit most imaging applications.
The DM IL LED Fluo inverted microscope platform (630-3113) for incident fluorescence with transmitted light LED illumination for bio-medicine applications, features a fixed stage on a 3-point support, coaxial double-knob for coarse and fine focus on both sides, a 4-fold revolving nosepiece with 25×0,75 objective thread, built-in fluorescence illumination axis with manual shutter, a fluorescence slider with three positions for filter blocks, a fluorescence port for exchangeable lamp houses (e.g. 50 W Hg# 100 W Hg), infinity optics with tube lens 1×, an interface for exchangeable observation tube and phototubes, and stepless, adjustable condenser holder, including collector, aperture diaphragm and filter holder. Illuminated ON/OFF switch. IMC interface. Power supply for TL LED illumination, including mains cable. The platform (630-3113) is included in both pre-configurations (630-3462 and 630-3463).
The DM IL LED inverted microscope platform (630-3112), with transmitted light LED illumination for bio-medicine applications, features a fixed stage on a 3-point support, a coaxial double-knob for coarse and fine focus on both sides, a 4-fold revolving nosepiece with 25×075 objective thread, infinity optics with tube lens 1×, an interface for exchangeable observation tube and phototubes, and stepless, adjustable condenser holder, including collector, aperture diaphragm and filter holder. Illuminated ON/OFF switch. IMC interface. Power supply including mains cable. The platform (630-3112) is included in pre-configurations (630-3464)
The DM IL Fluo Cellfactory inverted Microscope platform (630-3321) for large scale cell production ("Cellfactory") with 480 mm transmitted light column, fluorescence axis, and LED illumination.The Microscopes comes with a double-length (480 mm )TL-column, build in fluorescence illumination axis with manual shutter, fluorescence slider with 3 positions for filter blocks, fluorescence port for exchangeable lamp houses, a fixed stage on 3 point support, coaxial double knob for coarse and fine focus on both sides, a 4-fold revolving nosepiece with 25x0.75 objective thread, infinity optics with tube lens 1x, interface for exchangeable observation tube and phototubes, condenser holder stepless adjustable, including collector, aperture diaphragm and filter holder, illuminated ON/OFF switch and power supply including mains cable.
Certyfikaty: C-UL (Us) and CE.
Informacje do zamówienia: When ordering the DM IL LED with a camera please ensure that you select at least one of the three camera choices and the C-mount adapter. When selecting the LED light source please select all three items listed under the 'Fluorescence Light Source' heading. This includes the LED 3, the light guide coupler and the adapter sleeve. Optional filter cubes available. 2 year warranty.
Dodatkowe informacje: EU plug included. UK and CH plugs are sold separately.
DM IL LED Fluo pre-configuration package 1 without camera option includes microscope platform (630-3113); condenser S80 / 0.30; 4-fold lightring slider for S80, objective HI PLAN CY 10×/0,25 Ph1; binocular tube HC ILB; eyepiece HC PLAN 10×/20 BR.; eyepiece HC PLAN 10×/20 BR.M and dust cover for DM IL.
DM IL LED Fluo pre-configuration package 2 (camera upgradable) includes microscope platform (630-3113); condenser S80/0.30; 4-fold slider for light rings S80, objective HI PLAN CY 10×/0,25 Ph1; trinocular tube HC ILT; eyepiece HC PLAN 10×/20 BR.; eyepiece HC PLAN 10×/20 BR.M and dust cover for DM IL.
DM IL LED BF, PH pre-configuration package 3 (camera upgradable) includes microscope platform (630-3112); condenser S80 / 0.30; 4-fold lightring slider for S80, objective HI PLAN CY 10×/0,25 PH1, trinocular tube HC ILT; eyepiece HC PLAN 10×/20 BR.; eyepiece HC PLAN 10×/20 BR.M and dust cover for DM IL.
DM IL LED Fluo Cellfactory pre-configuration package 4 (camera upgradable) includes microscope platform (630-3321); condenser S80 / 0.30; 4-fold lightring slider for S80, objective HI PLAN CY 10×/0,25 PH1, trinocular tube HC ILT; eyepiece HC PLAN 10×/20 BR.; eyepiece HC PLAN 10×/20 BR.M and dust cover for DM IL.
Mniej informacji