Drukuj
VWR® VisiScope® 360, mikroskopy z zoomem stereo
Dostawca: VWR Collection
630-3090EA 3210 PLN
630-3090 630-3091 630-3085 630-3084 630-3087 630-3089 630-3086 630-3088
VWR® VisiScope® 360, mikroskopy z zoomem stereo
Mikroskopy
Choose from a range of binocular and trinocular stereo zoom microscope heads for either basic or advanced requirements.
  • Parfocal achromatic zoom
  • Greenough optical system
  • Available with or without illumination
  • Stable, slim modern stand or long overhanging stand for observation of particularly large specimens
  • Extended zoom range
Coarse focusing system (76 mm head holder, 32 mm pillar) with adjustable tension for standard stereo microscope requirements. Binocular head or trinocular head (split ratio: 50/50) feature 360° rotating on all stands and 45°Więcej informacji o produkcie