Drukuj
Ręczne mikroskopy cyfrowe, Dino-Lite Special Light, fluorescencja
Dostawca: Dino-Lite / IDCP B.V.
AM4115T-CFVW AM4115T-GFBW AM4115T-YFGW AM4115T-RFYW AM4115T-DFRW AM4515T4-GFBW
IDCPAM4115T-CFVWEA 4320 PLN
IDCPAM4115T-CFVW IDCPAM4115T-GFBW IDCPAM4115T-YFGW 630-2749 IDCPAM4115T-DFRW IDCPAM4515T4-GFBW
Ręczne mikroskopy cyfrowe, Dino-Lite Special Light, fluorescencja
Mikroskopy
These fluorescence microscopes are considered to be the world’s smallest fluorescence microscopes. Compared to the traditional fluorescence microscopes with the band-pass type of emission filters, the Dino-Lites long-pass emission filters provide visibility and sensitivity over a larger range of fluorescence wavelengths.
  • Visualise fluorescence from 400 nm to 620 nm
  • High speed USB 2.0 output
  • Exchangeable caps provide more flexibility
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable