Drukuj
VWR® Traceable®, barometr z rejestracją danych USB,
Dostawca: VWR Collection
620-2632EA 2120 PLN
620-2632
VWR® Traceable®, barometr z rejestracją danych USB,
Barometry
The VWR® Traceable® memory loc, USB, data logging barometer simultaneously monitors barometric pressure, temperature and humidity. This device can be used in 21 CFR 11 environments.
  • Memory capacity of 819150 temperature observations for barometric pressure, temperature and humidity
  • Raw data is locked and may not be cleared/changed on the base unit; unable to delete data from memory once stored
  • Calibration points: 25 °C for temperature, 50% RH for humidity, and 650, 850 and 1000 mbar for barometric pressure
  • Monitors barometric pressure, temperature and
Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable