Drukuj
VWR®, termometr trzpieniowy, kalibrowany, elektroniczny, stal nierdzewna, sonda 120 mm
Dostawca: VWR Collection
620-2200EA 337 PLN
620-2200
VWR®, termometr trzpieniowy, kalibrowany, elektroniczny, stal nierdzewna, sonda 120 mm
Termometry Termometry cyfrowe
Digital stem thermometer suitable for field studies, wet lab applications, wash-down areas, food testing and tanks.
  • Will read temperatures in liquids, semi-solids and gases
  • Stainless steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
  • Includes accredited calibration documentation that meets the requirements of ISO 17025
  • Individually serialised
Certyfikaty: Tested and calibrated in exclusive triple-accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448,01), against equipment whose calibration isWięcej informacji o produkcie