Drukuj
Bardzo dokładne termometry cyfrowe, TFN 520 and 530
Dostawca: EBRO
1340-5520 1340-5522
620-1460EA 1210 PLN
620-1460 620-1390
Bardzo dokładne termometry cyfrowe, TFN 520 and 530
Termometry Termometry cyfrowe
These thermometers can be used for measuring temperatures in air conditioning and ventilation applications, air and gases, liquids and for measuring surface temperatures, temperatures of tools for injection moulding processes and plastic materials.
  • Maximum/minimum values displayed
  • SMP, Lemo or Ex proof connections
  • 'HOLD' function
  • Automatic switch-off
  • Response times (t99) of three seconds (620-1461)
External thermocouple types: K, J, T, E probes.
Standard version IP 52, SMP versions IP 40.
Weight: 90 g
Standard version IP 52, SMP versions IP 40Więcej informacji o produkcie