Drukuj
Krioskopy, CryoStar
Dostawca: GERBER FUNKE
7150
614-2060EA 17170 PLN
614-2060
Krioskopy, CryoStar
Krioskopy
Quick and reliable measurement of the freezing point of milk. The cooling system provides quick operational readiness even at high ambient temperatures (up to 32 °C).
  • Easy to use, uncomplicated operation
  • Multi-language menu
  • Automatic calibration and settings history saved to non volatile memory
  • Software included for mapping the freezing curve on a PC
Certyfikaty: Reference measurement in accordance with DIN/ISO/IDF5764