Drukuj
Bottle-top dispensers, Varispenser® 2 and Varispenser® 2x
Dostawca: EPPENDORF
4966000.010 4966000.029 4966000.037 4966000.045 4966000.053 4966000.061 4967000.014 4967000.022 4967000.030 4967000.049 4967000.057 4967000.065
613-5915EA 1820 PLN
613-5915 613-5916 613-5917 613-5918 613-5919 613-5920 613-5921 613-5922 613-5923 613-5924 613-5925 613-5926
Bottle-top dispensers, Varispenser® 2 and Varispenser® 2x
Dozowniki butelkowe
Variable volume, autoclavable.
  • Discharge valve with safety ball closes and avoids fluid leakage when discharge tube is not mounted
  • Standard GL 45 thread on all sizes fits most common bottle threads
  • Screwable air vent cap allows easy assembly of optional drying tube to protect sensitive reagent against humidity or CO₂
  • Oval shaped dispenser housing allows safe gripping for transportation and adjustment
For laboratory tasks with aliquots of aggressive solutions like lyes, acids, bases or solvents taken from large supply and reagent bottles, bottle topWięcej informacji o produkcie