Drukuj
Dozowniki butelkowe, FORTUNA OPTIFIX®
Dostawca: Poulten & Graf
101.08027 101.08033 101.08038 101.08044 101.08048 101.08051 101.08053 101.08056 101.08058 101.08127 101.08133 101.08138 101.08144 101.08148 101.08151 101.08153 101.08156 101.08158 101.10638 101.10644 101.10727 101.10733 101.10738 101.10744 101.10748 101.10751 101.10748S 101.10738S 101.10744S 101.10633 101.10733S 101.10751S 101.10727S
612-2112EA 1920 PLN
612-2112 612-2113 612-2114 612-2115 612-2116 612-2117 612-2118 612-2119 612-2120 612-2142 612-2143 612-2144 612-2145 612-2146 612-2147 612-2148 612-2149 612-2150 612-2194 612-2195 612-2172 612-2173 612-2174 612-2175 612-2176 612-2177 613-3134 613-3132 613-3133 613-3131 615-0008 615-0009 615-0007
Dozowniki butelkowe, FORTUNA OPTIFIX®
Dozowniki butelkowe
This range of four models is suitable for many applications. A discharge tube is screwed on the valve block. The BASIC and SOLVENT models are autoclavable.
  • High quality materials and robust design for a long life-span
  • Precise dispensing: Innovative volume setting system with a fast and easy setting and a fine adjustment
  • Colour-code: Each model provides a special colour on the finger protection cap for a quick and safe identification
  • Safe and efficient dispensing of dangerous chemicals
  • No metal spring in the OPTIFIX® dispenser, no contamination of the
Więcej informacji o produkcie