Drukuj
Rozdzielacz do kapilar, b.safe
Dostawca: Bohlender
M511-01
590-0195EA 257 PLN
590-0195
Rozdzielacz do kapilar, b.safe
Okucia i złącza
Distributor for connection of capillary tubing. With PFA Fittings for capillaries with Ext.Ø 3,2 mm (thread UNF ¹/₄").
  • Excellent chemical resistance
  • Available for a wide range of hose diameters
  • The conical shape gets the connection tight and leak-free
Adaptor to enlarge the number of capillary connectors on b.safe Caps.