Drukuj
VWR®, Kliniczne 4-drożne statywy na probówki
Dostawca: VWR Collection
525-1486EA 627 PLN
525-1486 525-1484 525-1485 525-1487 525-1488
VWR®, Kliniczne 4-drożne statywy na probówki
Statywy do probówek Tube Racks
These tube racks have a six way connection to create various configurations and are designed to hold round, flat and conical blood collection and culture tubes.
  • Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
  • Molded alphanumeric identification for quick location and retrieval of tubes
  • Made from polypropylene that is chemical resistant and easy to clean
  • Racks can be used in water baths and placed in freezer
  • Accommodates all brands of 13 and 17 mm tubes
They can be used as standalone unit or infinite units canWięcej informacji o produkcie