Drukuj
Wirówki o wysokiej wydajności, Avanti™ JXN-26
Dostawca: Beckman Coulter
B34182 B38618
521-4188EA 0 PLN
521-4188 521-4192
Wirówki o wysokiej wydajności, Avanti™ JXN-26
Wirówki Wirówki podłogowe
Experience simplified high-throughput sample processing and more control with the JXN-26 centrifuge with powerful high speed features, low heat output, imbalance tolerance and quiet performance. The JXN-26 has a maximum capacity of 6,0 L and 2,6×10⁴ min⁻¹ (82000×g). The Friction Reduction System (FRS) means the JNX-26 runs quicker, quieter and cooler, with shorter cycle times with high-torque because of the exclusive Switched Reluctance (SR) drive. Stores up-to 1000 user-defined programs.
  • MobileFuge app via Apple iOS and Android, adds the ability to access
Więcej informacji o produkcie