Drukuj
Wirnik, Avanti JS-4.3
Dostawca: Beckman Coulter
362734
521-3989EA 0 PLN
521-3989
Wirnik, Avanti JS-4.3
Wirówki Wirniki
Four-place rotor with anodised aluminium buckets designed for use with modular disk adapters to accommodate a variety of tubes and other adapters and blood bag cups. Can be used with Beckman Coulter aerosolve canisters which are designed to provide added protection from aerosols. Rotor buckets are interchangeable with microplate carriers.