Drukuj
3-częściowe lejki filtracyjne, Whatman™
Dostawca: Whatman products (Cytiva)
1950-002 1950-004 1950-007 1950-009 1950-012 1950-017 1950-027
516-0605EA 1280 PLN
516-0605 516-0606 516-0607 516-0608 516-0609 516-0610 516-0611
3-częściowe lejki filtracyjne, Whatman™
Lejki, rozdzielacze i wkraplacze Lejki filtracyjne Lejki filtracyjne
Whatman 3-piece filter funnels from Cytiva are ideal for quick and easy filtration using high-efficiency glass microfiber filters.
  • Chemically resistant - 3-piece construction consists of glass cylinder and base with a polymeric membrane support
  • Support options - Provided with acrylic support and optional polypropylene or PTFE supports available separately for some sizes
  • Secure sealing - Clamps at edges to prevent peripheral loss due to fluid bypass around the edge of the filter
  • Eliminate carry-over - Simplicity of design facilitates easy and thorough
Więcej informacji o produkcie