Drukuj
Filtry membranowe z siatką
Dostawca: Sartorius
13006-47-ACR 13006-50-ACR 13806-47-ACN 13806-47-R 13806-50-R 13906-50-ACR 13906-50-R 11403-25-N 11403-47-ACR 11404-37-N 11404-47-ACN 11404-47-ACR 11404-50-ACN 11405-47-ACN 11406-50-ACR 11406-82-K 11407-47-ACR 11407-50-ACR 11407-50-N 13004-47-ACN 13004-50-N 13005-47-N 13005-50-ACR 11403-47-ACN 11403-47-N 11404-47-N 11404-50-N 11406-25-N 11406-47-ACN 11406-47-N 11406-50-ACN 11406-50-N 11407-47-ACN 11407-47-N 11407-50-ACN 13005-50-ACN 13005-50-N 13006-47-ACN 13006-47-N 13006-50-ACN 13006-50-N 13806-47-N 13806-50-ACN 13806-50-ACR 13806-50-N 13906-47-N 11406-47-ACR 13004-47-N 13004-47-ACR 13005-47-ACN 11404-25-N 13906-50-ACN 13906-47-ACR 13906-47-ACN 13806-47-ACR 13006-25-N 114H6-47-ACR 114H6-50-ACR 139H6-47-ACR 13006-47-R 114H6-47-ACN 139H6-50-ACN 139H6-47-ACN 11406-47-R 11407-47-R 130H6-50-ACR 130H6-47-ACR 130H6-47-ACN
515-0223EA 4750 PLN
515-0223 515-0226 514-7006 515-0232 512-5074 515-0236 515-0237 515-0190 515-0191 515-0194 611-1155 515-0195 515-0196 514-0575 515-0183 SART11406-82-K 514-0576 515-0184 515-0213 611-1156 515-0219 515-0220 515-0221 512-5011 512-5010 512-5012 512-5023 512-5018 514-7001 512-5014 514-7003 512-5024 512-5017 512-5016 512-5027 512-5052 512-5051 512-5035 512-5034 514-7005 512-5054 512-5071 512-5073 512-5075 512-5072 512-5081 611-0679 515-0217 515-0216 611-0681 515-0193 514-7009 611-0685 514-7008 611-0684 515-0222 515-0185 515-0215 515-0239 515-0224 515-0214 515-0240 515-0238 515-0199 515-0212 515-0230 515-0228 515-0227
Filtry membranowe z siatką
Filtry Filtry membranowe
Cellulose nitrate hydrophilic, gridded membrane filters for colony counting, particle testing and microscopy, with outstanding recovery rates for microorganisms.
Membranes with 0,45 µm pore size are tested and released according to ISO 7704.