Drukuj
Analytical Membrane Bands
Dostawca: Thermo Fisher Scientific
DS0600-0102 DS0600-0108 DS0600-0145 DS0600-0202 DS0600-0208 DS0600-0245
514-1430EA 3210 PLN
514-1430 514-1431 514-1432 514-1433 514-1434 514-1435
Analytical Membrane Bands
Filtry Filtry membranowe
Analytical membrane bands are perfect for both microbial quality assurance (QA) and quality control (QC) applications where membrane dispensers are used.
  • Membranes made of MCE material
  • Bands are numbered to indicate how many membranes remain to be dispensed
  • Membranes are gridded to improve accuracy and save time
Compatible with most common dispensers on the market, including MilliporeSigma™ and Sartorius™, giving added flexibility and security of supply. Boxes feature two marked orientations and tear-away perforations with a thumb-tab for quick andWięcej informacji o produkcie