Drukuj
Tygle filtracyjne, VitraPOR®
Dostawca: ROBU GLASFILTER
20082 20083 20084 20151 20152 20153 20154 20301 20302 20304 20305 20501 20502 20503 20504 20505 20155 20080 20081 20150 20300 20500 20085 2050C 2030C 2030F 20303 2050F
511-2102EA 76.1 PLN
511-2102 511-2103 511-2104 511-2201 511-2202 511-2203 511-2204 511-2301 511-2302 511-2304 511-2305 511-2401 511-2402 511-2403 511-2404 511-2405 511-0360 511-0358 511-2101 511-0359 511-0249 511-0361 511-0248 ROBU2050C ROBU2030C ROBU2030F 511-0273 ROBU2050F
Tygle filtracyjne, VitraPOR®
Tygle
Filtration crucibles made from borosilicate glass 3.3 with integrated sintered glass filter plate.
Certyfikaty: Porosity 0 to 5 comply with ISO 4793-80. Porosity C (coarse) and F (fine) comply with ASTM E128-94.