Drukuj
Autoklawy, poziome i pionowe, sterowanie półautomatyczne, seria ML/MLV
Dostawca: TUTTNAUER
3870ML 3870MLV
481-2067EA 65130 PLN
481-2067 481-2070
Autoklawy, poziome i pionowe, sterowanie półautomatyczne, seria ML/MLV
Autoklawy Autoklawy (sterylizatory parowe)
Tuttnauer's ML laboratory line of semi-automatic, electro-mechanically controlled, vertical steam sterilizers provide safe, economical and effective sterilization. The semi-automatic laboratory autoclave series is an affordable sterilizer for laboratories that do not want to compromise on quality, safety and reliability and need to sterilize loads with low sensitivity. The semi-automatic series is available in vertical or benchtop models and in chamber sizes ranging from 23 to 160 litres. The ML models have a semi-automatic control panel, ML models have a manual control panel. The chamber isWięcej informacji o produkcie