Drukuj
Statywy dla 48 próbówek do przechowywania, Nunc™
Dostawca: Thermo Fisher Scientific
331830 331835 331830-BR 331835-BR
479-1195EA 523 PLN
479-1195 479-1867 479-2492 479-2493
Statywy dla 48 próbówek do przechowywania, Nunc™
Statywy do probówek Statywy na vialki kriogeniczne
Use versatile Thermo Scientific™ Nunc™ 1,8 ml Externally-threaded universal tubes inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes or non-coded.
  • Barcoded rack
  • Sterile
  • Code type: 2D, linear and human readable
  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cryostorage boxes made for 10×10 arrays
  • External threads limit contamination
  • Automation-pickable in
Więcej informacji o produkcie