Drukuj

Iron, plasma standard solution, Fe 10000 µg/ml, Specpure®

Dostawca: Thermo Fisher Scientific

47221.AD 47221.AE 47221.AP
47221.ADEA 785 PLN
47221.AD 47221.AE 47221.AP
Iron, plasma standard solution, Fe 10000 µg/ml, Specpure®
Standardy Wzorce kationowe Iron Standards

Molecular Formula: (Fe(NO₃)₃).9H₂O in 5% HNO₃
Physical Form: Liquid
Odour: Acidic
UN No.: UN3265
MDL No.: MFCD00010999

Zobacz ceny

SPECYFIKACJE

Environmentally Preferable
Nr. MDL MFCD00010999

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR