Drukuj
Eksykatory, mini próżniowe, Bel-Art
Dostawca: Bel-Art Products, a Part of SP
F410990000
467-2245EA 1100 PLN
467-2245
Eksykatory, mini próżniowe, Bel-Art
Eksykatory Eksykatory z tworzyw sztucznych
Desiccators with top made of clear polycarbonate, for easy viewing of contents, and white polypropylene bottom. The small size of this desiccator allows quick vacuum creation for rapid, repeated degassing of multiple samples.
  • Vacuum rating of 740 mm Hg
  • Stopcock with 2 mm PTFE plug accepts tubing with ¼ to ³/₈" int.Ø
Peripheral flange around the bottom fits in to two snap locks and seals the two halves against a Buna-N O-ring.