Drukuj
Suszarki, seria Heratherm® Advanced protocol, Thermo Fisher Scientific
Dostawca: Thermo Fisher Scientific
51028153 51028152 51028151 51028142 51028143 51028144 51028715 51028709 51028711 51028713 51028716 51028712
466-0275EA 27520 PLN
466-0275 466-0274 466-0273 466-0267 466-0268 466-0269 466-0277 466-0270 466-0271 466-0276 466-0278 466-0272
Suszarki, seria Heratherm® Advanced protocol, Thermo Fisher Scientific
Piece i suszarki Piece i suszarki standardowe
Heating and drying ovens with coated or stainless steel exterior (SS models). The timer allows the oven to be turned on or off after a pre-set time, with a choice between weekly, hourly or real time settings. Units with natural convection are ideal for gentle drying and heating, to protect delicate samples. Units with forced convection have adjustable fan speed and are suitable for faster drying and heating, and offer superior temperature uniformity.
  • Automatic over-temperature alarm system
  • Large, easy to read vacuum fluorescent display and microprocessor
Więcej informacji o produkcie