Drukuj
VWR®, Sulphur standard solutions for ICP-MS, ARISTAR®
Dostawca: VWR Chemicals
456922XEA 977 PLN
456922X 85594.180
VWR®, Sulphur standard solutions for ICP-MS, ARISTAR®
Standardy Wzorce kationowe Sulphur Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0.5% (typical)
  • Delivered with complete certificate of analysis
Certyfikaty: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable