Drukuj
Mieszadło magnetyczne, Variomag Power Direct
Dostawca: THERMO H + P LABORTECHNIK
50098760
442-0162EA 7200 PLN
442-0162
Mieszadło magnetyczne, Variomag Power Direct
Mieszadła Mieszadła magnetyczne
Magnetic stirrer with compact, durable design. Unit has a powerful, far-reaching magnetic field, which enables efficient stirring, even when there is some distance between stirrer and stirring bar.
  • Wear-free and maintenance free (as unit has no moving parts such as motors, belts or mechanical bearings)
  • Safe, low operation voltage; automatic start-up for safe stirrer acceleration; quick stop function stops stirring in two seconds to prevent vessels breaking
  • Memory function stores the most recently used settings, even when the device is unplugged for a long
Więcej informacji o produkcie