Drukuj
Worki do mikserów laboratoryjnych, Stomacher
Dostawca: Seward (FermionX)
BA6040 BA6141 BA6041/CLR BA6141/STR BA6141/CLR BA6041/5/500 BA6042 BA6141/5/500 BA6041/STR BA6041 BA6040/CLR BA6040/CLR/STR BA6040/DBL/STR BA6040/STR BA6042/HD BA6041/LSTR
432-0017EA 1020 PLN
432-0017 432-3121 432-3123 432-3124 432-3125 431-0130 432-3131 432-0025 432-3119 432-3126 432-0271 432-0272 432-3118 432-0273 SEWABA6042/HD SEWABA6041/LSTR
Worki do mikserów laboratoryjnych, Stomacher
Worki Worki do homogenizatorów
Stomacher® bags are all manufactured from virgin polymer granule extruded into a tube. The only seal is at the base. All bags are batch tested and in-process monitored during manufacturing on Stomacher® machines. Bags are tested against a wide range of parameters that include open-ability, transparency, and flexibility as well as dimensions and performance. Stomacher® bags come in a range of formats to suit all applications.
  • Manufactured from high-grade, food quality polythene
  • Minimum 65 µm film thickness provides high impact strength
  • Gamma irradiated at 25
Więcej informacji o produkcie
Zobacz ceny
SPECYFIKACJE
Environmentally Preferable